Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POF1B Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548641

Catalog No. PA548641

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a key role in the organization of epithelial monolayers by regulating the actin cytoskeleton. May be involved in ovary development.
Specifications

Specifications

POF1B
Polyclonal
PBS with 0.09% sodium azide
Q8K4L4
Protein POF1B, Premature ovarian failure protein 1B, POF1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
69693
Western Blot
Unconjugated
Pof1b
Liquid
POF1B
KLH conjugated synthetic peptide between 337-366 amino acids from the Central region of human POF1B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit