Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POFUT2 Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: OTI3A12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273512R
Description
POFUT2 Monoclonal specifically detects POFUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
POFUT2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POFUT2 | |
Full length recombinant protein of human POFUT2 (NP_598368) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI3A12 | |
Western Blot, Flow Cytometry | |
C21orf80, chromosome 21 open reading frame 80, FUT13EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 2, KIAA0958, O-FucT-2, Peptide-O-fucosyltransferase 2, protein O-fucosyltransferase 2 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
23275.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title