Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POGK Mouse anti-Human, Mouse, Rat, Monkey, DyLight 488, Clone: OTI6D5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273513G
Description
POGK Monoclonal specifically detects POGK in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POGK | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POGK | |
Full length recombinant protein of human POGK (NP_060012) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI6D5 | |
Western Blot, Immunohistochemistry | |
BASS2, KIAA1513, KIAA15131, KRAB box domain containing 2, KRBOX2, LST003, pogo transposable element with KRAB domain | |
Mouse | |
Affinity Purified | |
Apoptosis | |
57645.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title