Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POGK Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | POGK |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180337
|
Novus Biologicals
NBP180337 |
100 μL |
Each for $436.00
|
|
NBP18033720
|
Novus Biologicals
NBP18033720UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
POGK Polyclonal specifically detects POGK in Human samples. It is validated for Western Blot.Specifications
POGK | |
Polyclonal | |
Rabbit | |
Apoptosis | |
BASS2, KIAA1513, KIAA15131, KRAB box domain containing 2, KRBOX2, LST003, pogo transposable element with KRAB domain | |
POGK | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_060012 | |
57645 | |
Synthetic peptide directed towards the middle region of human POGK. Peptide sequence SSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAES. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title