Learn More
Thermo Scientific™ Point of Use Filter Inserts
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters.
$105.00 - $210.00
Catalog Number | Mfr. No. | Description | Flow Rate | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Flow Rate | For Use With (Equipment) | Price | Quantity & Availability | |||
10-451-113
|
Thermo Scientific™
06.5101 |
Prefilter 1µm (10“) | 1000 L/hr. (at D p = 0.2 bar) | Smart2Pure |
Each for $105.00
|
|
10-451-114
|
Thermo Scientific™
06.5201 |
Prefilter 5µm+activated carbon (10“) | 750 L/hr. (at D p = 0.2 bar) | LabTower EDI, LabTower RO |
Each for $210.00
|
|
Description
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters. Thermo Scientific™ Point of Use Filter Inserts include 10- and 20-inch pre-filters, final filters and sterile filters for Point of Use systems.
- Protect installations and systems from undissolved substances in water
- Protect systems from undissolved substances in water
- Usually used for final filtration
- For final filtration and delivery of organic-free water
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.