Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Alkaline Phosphatase Reagents

For quantitative determination of Alkaline Phosphatase in human serum

$303.39

Specifications

Certifications/Compliance CE Marked
For Use With (Application) Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
pH 10.45
Product Type Alkaline Phosphatase reagent kit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Formulation Price Quantity & Availability  
23-666-079
View Documents
Pointe Scientific
HA916492
4 x 100mL (R1); 4 x 23mL (R2) R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives
Pack of 8 for $303.39
Add to cart
 
Description

Description

HA716-246 is for Hitachi 704, 717. HA916-492 is for Hitachi 911, 912, 914.

  • Format: Dry Powder
  • Wavelength: 405nm
  • Linearity: 800 U/L
  • Expected Values: Adults 35-123 U/L, Children have a higher normal value
  • Reconstituted Stability: 7 days RT, 60 days 2°-8°C

Specifications

Specifications

CE Marked
Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
10.45
Alkaline Phosphatase reagent kit
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit