Learn More
Pointe Scientific Alkaline Phosphatase Reagents
For quantitative determination of Alkaline Phosphatase in human serum
Manufacturer:  Pointe Scientific A7505300
 View more versions of this product
Catalog No. 23-666-083
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
10.45
Alkaline Phosphatase reagent kit
Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
Alkaline Phosphatase reagent p-Nitrophenylphosphate 10mM, Magnesium Ions 1mM, Buffer pH 10.1 ±0.1, activator and binder
20 x 15mL