Pointe Scientific Alkaline Phosphatase Reagents

For quantitative determination of Alkaline Phosphatase in human serum

Manufacturer: Pointe ScientificSupplier Diversity Partner A7505-400

 View all options for this product

Catalog No. 23-666-086


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
    Format Dry powder
    Formulation Alkaline Phosphatase reagent p-Nitrophenylphosphate 10mM, Magnesium Ions 1mM, Buffer pH 10.1 ±0.1, activator and binder
    pH 10.45
    Quantity 8 x 50mL
    Product Type Alkaline Phosphatase reagent kit

    HA716-246 is for Hitachi 704, 717. HA916-492 is for Hitachi 911, 912, 914.

    • Format: Dry Powder
    • Wavelength: 405nm
    • Linearity: 800 U/L
    • Expected Values: Adults 35-123 U/L, Children have a higher normal value
    • Reconstituted Stability: 7 days RT, 60 days 2°-8°C

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit