Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific ALT Reagents for Hitachi Analyzers
For quantitative determination of alanine aminotransferase in serum on Hitachi analyzers
Supplier: Pointe Scientific HA926610
This item is not returnable.
View return policy
Description
HA726-305 is for Hitachi 704. HA926-610 is for Hitachi 911, 912, 914. HA426-R1 and HA426-R2 for Hitachi 747.
- Method: Modified IFCC, Kinetic
- Format: Liquid
- Linearity: 0-500 U/L
- Expected Values: 4-36 U/L
- Storage Temperature: 2° to 8°C
- Reconstituted Stability: Ready to use liquid
Specifications
CE Marked | |
R1 and R2 contains: L-alanine 500mM, alpha-ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM, pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers | |
Hitachi 717 analyzers | |
Liquid |
ALT Reagent Kit | |
Quantitative determination of Alanine Aminotransferase in Serum | |
5 x 100mL R1, 2 x 50mL R2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction