Pointe Scientific ALT (SGPT) Dry Reagent Sets

Manufacturer: Pointe ScientificSupplier Diversity Partner A7525300

 View all options for this product

Catalog No. 23-666-092


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Alanine Aminotransferase in Serum, in-vitro diagnostic use only
    Format Powder
    Formulation alpha-ketoglutaric Acid 13mM, L-alanine >400mM, NADH >0.2mM, LDH >1200U/L, Tris Buffer pH 7.5 ± 0.1, non-reactive fillers and Stabilizers
    Quantity 300mL
    Product Type ALT (SGPT) dry reagent set

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit