Learn More
Pointe Scientific ALT (SGPT) Dry Reagent Sets
Manufacturer:  Pointe Scientific A7525400
 View more versions of this product
Catalog No. 23-666-094
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
CE Marked
Powder
400mL
Quantitative determination of Alanine Aminotransferase in Serum, in-vitro diagnostic use only
alpha-ketoglutaric Acid 13mM, L-alanine >400mM, NADH >0.2mM, LDH >1200U/L, Tris Buffer pH 7.5 ± 0.1, non-reactive fillers and Stabilizers
ALT (SGPT) dry reagent set