Pointe Scientific AST (SGOT) Dry Reagent Sets

For quantitative determination of aspartate aminotransferase in serum

Manufacturer: Pointe Scientific A7560150

 View more versions of this product

Catalog No. 23-666-123

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Aspartate Aminotransferase in Serum, in-vitro diagnostic use only
    Format Powder
    Formulation alpha-ketoglutarate 12mM, L-aspartic Acid >150mM, NADH >0.2mM, LDH >800U/L, MDH >600U/L, Tris Buffer pH 7.8 ±0.1, non-reactive fillers and Stabilizers
    Quantity 150mL
    Product Type AST (SGOT) dry reagent set

    • Method: optimized Tris buffer, kinetic
    • Reconstituted stability: 48 hours at room temperature, 30 days at 2° to 8°C
    • Linearity: 500U/L
    • Wavelength: 340nm
    • Expected values: up to 40U/L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit