Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific autoHDL Cholesterol Reagents
Specifications
Certifications/Compliance | CE Marked |
---|---|
Format | Liquid |
For Use With (Application) | Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma |
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||
23-666-134
|
Pointe Scientific
HH445R1 |
5 x 250 R1mL | R1: alpha-cyclodextrin 0.5mM, Dextran Sulfate 0.5g/L, Magnesium Chloride 2mM, HSDA 0.3g/L, Buffer pH 7 ± 0.1, preservative | Hitachi™ 747 analyzer | AutoHDL™ Cholesterol R1 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-135
|
Pointe Scientific
HH445R2 |
3 x 150 R2mL | R2: POD >15000U/L, PEG-CO >5000U/L, PEG-CE >800U/L, 4-aminoantipyrene 0.5g/L, Buffer pH 7 ± 0.1, Surfactant, preservative | Hitachi™ 747 analyzer | AutoHDL™ Cholesterol R2 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-133
|
Pointe Scientific
HH945240 |
6 x 30mL R1, 3 x 20mL R2 | Hitachi™ 911, 912, 914 analyzer | AutoHDL™ Cholesterol reagent kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Requires autoHDL/LDL Calibrator (23-666-140). HH945-240 is for Hitachi 911, 912, 914. HH445-R1 and HH4450R2 are for Hitachi 747.
Specifications
CE Marked | |
Liquid | |
Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title