Learn More
Pointe Scientific BUN (Urea Nitrogen) UV Reagents
Manufacturer:  Pointe Scientific B7550-75
 View more versions of this product
Catalog No. 23-666-165
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
CE Marked
Powder
75mL
Quantitative determination of Urea Nitrogen in Serum, in-vitro diagnostic use only
NADH 0.3mM, Urease 1,500U/L, Glutamate Dehydrogenase >1500U/L, alpha-Ketoglutarate 4mM, pH 8.2 ± 0.1 Buffer, activators and non-reactive Stabilizers
Urea Nitrogen (BUN) UV reagent kit