Learn More
Pointe Scientific™ Calcium Dry Powder Reagents
Quantify calcium levels in serum
Manufacturer:  Pointe Scientific C7508-300
 View more versions of this product
Catalog No. 23-666-174
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Requires : Requires Calibrator/Standard
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
20 x 15mL
570nm
Quantitative determination of Calcium in Serum
Calcium reagent : o-Cresolphthalein Complexone 0.1mM, 8-Hydroxyquinoline Sulfonate 8.88mM, Buffer 100mM, pH 10.1 ±0.1, nonreactive stabilizers and fillers with Sodium Azide 0.01% as preservative
20 × 15mL
Calcium reagent kit