Pointe Scientific™ Calcium Dry Powder Reagents

Quantify calcium levels in serum

Manufacturer: Pointe Scientific C7508400

 View more versions of this product

Catalog No. 23-666-176

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


Required Accessories : Requires Calibrator/Standard

    Specifications

    Product Type Calcium reagent kit
    For Use With (Application) Quantitative determination of Calcium in Serum
    Format Dry powder
    Includes Calcium reagent : o-Cresolphthalein Complexone 0.1mM, 8-Hydroxyquinoline Sulfonate 8.88mM, Buffer 100mM, pH 10.1 ±0.1, nonreactive stabilizers and fillers with Sodium Azide 0.01% as preservative
    Quantity 8 x 50mL
    Size 8 × 50mL
    Wavelength 570nm
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit