Learn More
Pointe Scientific™ Cholesterol Dry Powder Reagents
Quantify the total cholesterol in serum
Manufacturer:  Pointe Scientific HC9101000
 View more versions of this product
Catalog No. 23-666-197
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CE Marked
Hitachi™ 717, 911, 912, 914 analyzer
4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives
100mL
Quantitative determination of total Cholesterol in Serum
Liquid
6.7
Cholesterol reagent set