Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Cholesterol Dry Powder Reagents
Quantify the total cholesterol in serum
Specifications
Certifications/Compliance | CE Marked |
---|---|
For Use With (Application) | Quantitative determination of total Cholesterol in Serum |
Includes: 4-Aminoantipyrine 0.6mM, Sodium Cholate 8.0mM, Cholesterol Esterase ≥150u/L, Cholesterol Oxidase ≥200u/L, Horseradish Peroxidase 1000u/L, ρ-Hydroxybenzene Sulfonate 20mM, Surfactant, Buffer, pH 7.2, non-reactive stabilizers and fillers with Sodium Azide (.01%)
Catalog Number | Mfr. No. | Quantity | Format | Formulation | pH | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Format | Formulation | pH | Price | Quantity & Availability | ||
23-666-196
|
Pointe Scientific
HC4103000 |
600mL | Liquid | 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives | 6.7 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-194
|
Pointe Scientific
HC710600 |
50mL | Liquid | 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives | 6.7 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-205
|
Pointe Scientific
C75092L |
500mL | Dry Powder | 4-Aminoantipyrine 0.6mM, Sodium Cholate 8mM, Cholesterol Esterase <150μ/L, Cholesterol Oxidase <200μ/L, Horseradish Peroxidase 1000μ/L, p-Hydroxybenzene Sulfonate 20mM, Surfactants, Buffer pH 7.2, non- | 7.2 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Method: Enzymatic, Endpoint
- Format: Dry powder
- Wavelength: 520nm
- Linearity: 500mg/dL
- Expected Values: <200mg/dL (desirable cholesterol); 200-239mg/dL (borderline-high); >240mg/dL (high)
- Reconstituted stability: 60 days at 2 to 8°C
For the quantitative determination of Total Cholesterol in serum
Specifications
CE Marked | |
Quantitative determination of total Cholesterol in Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title