Pointe Scientific™ Cholesterol Dry Powder Reagents

Quantify the total cholesterol in serum

Manufacturer: Pointe ScientificSupplier Diversity Partner C7509-1L

 View all options for this product

Catalog No. 23-666-204


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of total Cholesterol in Serum
    Format Dry powder
    Formulation 4-Aminoantipyrine 0.6mM, Sodium Cholate 8mM, Cholesterol Esterase <150μ/L, Cholesterol Oxidase <200μ/L, Horseradish Peroxidase 1000μ/L, p-Hydroxybenzene Sulfonate 20mM, Surfactants, Buffer pH 7.2, non-
    pH 7.2
    Quantity 250mL
    Size 4 × 250mL (8.5 oz.)
    Product Type Cholesterol reagent kit

    • Method: Enzymatic, Endpoint
    • Format: Dry powder
    • Wavelength: 520nm
    • Linearity: 500mg/dL
    • Expected Values: <200mg/dL (desirable cholesterol); 200-239mg/dL (borderline-high); >240mg/dL (high)
    • Reconstituted stability: 60 days at 2 to 8°C

    For the quantitative determination of Total Cholesterol in serum

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit