Pointe Scientific™ Cholesterol Dry Powder Reagents

Quantify the total cholesterol in serum

Manufacturer: Pointe Scientific C7509400

 View more versions of this product

Catalog No. 23-666-207

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Cholesterol reagent kit
    For Use With (Application) Quantitative determination of total Cholesterol in Serum
    Format Dry powder
    Formulation 4-Aminoantipyrine 0.6mM, Sodium Cholate 8mM, Cholesterol Esterase <150μ/L, Cholesterol Oxidase <200μ/L, Horseradish Peroxidase 1000μ/L, p-Hydroxybenzene Sulfonate 20mM, Surfactants, Buffer pH 7.2, non-
    pH 7.2
    Quantity 50mL
    Size 8 × 50mL (1.7 oz.)
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit