Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific CK Reagents
$638.60
Specifications
Certifications/Compliance | CE Marked |
---|---|
Format | Liquid |
For Use With (Application) | Quantitative determination of Creatine Kinase activity in Serum |
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Formulation | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||
23-666-213
|
Pointe Scientific
HC922250 |
2 x 100mL R1, 2 x 25mL R2 | CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A | Hitachi™ 747 analyzer | Creatine Kinase reagent kit |
Pack of 4 for $638.60
|
|
23-666-215
|
Pointe Scientific
HC422R2 |
3 x 150 R2mL | CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), ADP 2mM/L, AMP 5mM/L, Diadensosine pentaphosphate 10mM/L, Creatine Phosphate 30mM/L, G6PDH (Baker's Yeast) 1.5KU/L, EDTA 2mM/L | Hitachi™ 747 analyzer | Creatine Kinase R2 kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-214
|
Pointe Scientific
HC422R1 |
4 x 500 R1mL | CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine 20mM/L | Hitachi™ 747 analyzer | Creatine Kinase R1 kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Specifications
CE Marked | |
Liquid | |
Quantitative determination of Creatine Kinase activity in Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title