Learn More
Pointe Scientific Direct Hemoglobin A1c (HbA1c)
Single Channel Method
Catalog Number | Mfr. No. | Quantity | For Use With (Application) | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | For Use With (Application) | Product Type | Price | Quantity & Availability | |||
22-666-102
|
Pointe Scientific
H7541CAL |
4 x 0.5mL | Calibrating results in the quantitative determination of Human hemoglobin A1c in Blood by automated immunoassay, for in-vitro diagnostic use only | Hemoglobin A1c calibrator set |
Pack for $176.94
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
22-666-103
|
Pointe Scientific
H7541CTL |
4 x 0.5mL | Monitoring accuracy and precision in the quantitative determination of Human hemoglobin A1c in Blood by automated immunoassay, for in-vitro diagnostic use only | HbA1c control |
Pack for $152.44
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
22-666-104
|
Pointe Scientific
H7541LYS |
1 x 500mL | Determination of HbA1c is most commonly performed for the evaluation of glycemic control in diabetes mellitus | HbA1c lyse |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
- A latex-enhanced immunoturbidimetric assay for the automated determination of HbA1c
- For use with routine chemistry analyzers
- Calibrators referenced to NGSP value (assigned HbA1c frozen whole blood)
- Normal and elevated controls are available
- Reagents are liquid stable for cost-effective use of the entire kit
Specifications
Routine chemistry analyzer |