Pointe Scientific Direct Hemoglobin A1c (HbA1c)
Single Channel Method
Manufacturer: Pointe Scientific H7541120
 View more versions of this product
Catalog No. 22-666-101
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type HbA1c kit
    For Use With (Application) Quantitative determination of hemoglobin A1c in Human Blood
    For Use With (Equipment) Routine chemistry analyzer
    Linearity 2 to 16mg/dL
    Quantity 450 Tests
  • Description & Specifications
Related Products
Product Content Feedback