Learn More
Pointe Scientific General Chemistry Calibrators
Liquid-stable, serum-based calibrator sets contain both Total and Direct Bilirubin values
Manufacturer:  Pointe Scientific F7546CAL
 View more versions of this product
Catalog No. 23-666-254
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fructosamine Calibrator
Liquid
CE
1 x 2mL
Clear, Slightly Yellow
3mM/L
2° to 8°C