Learn More
Pointe Scientific General Chemistry Calibrators
Liquid-stable, serum-based calibrator sets contain both Total and Direct Bilirubin values
Manufacturer:  Pointe Scientific B7538-CAL-3
 View more versions of this product
Catalog No. 23-666-146
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Bilirubin Calibrator Set
Liquid
For use as a calibrator in the determination of direct and total bilirubin assays
2° to 8°C
Clear and Yellow-Orange
0.005g/dL
CE
1 x 3mL