Pointe Scientific General Chemistry Controls

Manufacturer: Pointe Scientific H75066

 View more versions of this product

Catalog No. 23-666-307

  Check Availability
Add to Cart

    Specifications

    Product Type Hemoglobin control set
    Format Lyophilized
    For Use With (Application) In-vitro diagnostic use, Human hemoglobin controls for the cyanmethemoglobin hemoglobin procedure
    Certifications/Compliance CE
    Storage Requirements 2° to 8°C
    Quantity 3 x 2mL,3 x 2mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit