Learn More
Pointe Scientific General Chemistry Controls
Manufacturer:  Pointe Scientific G75402
 View more versions of this product
Catalog No. 23-666-297
Add to cart
This item is not returnable. View return policy
Description
Description
  • Human Serum Based (Lyophilized): Chemistry Control (Level 1)(23-666-187), Chemistry Control (Level II)(23-666-188), and Chemistry Control (Level 1 and II)(NC9896982)
  • Hemoglobin Control Set (23-666-307) contains Human Erythrocytes (Lyophilized). Contains three levels of hemoglobin.
  • Lipid Control Set (23-666-359) is Human Serum Based (Lyophilized). Expected Values: The lipid values were determined by repetitive assays of the listed methods. Values listed are targets only.
  • Measurements using other reagents and instrument systems may give different results
  • Listed ranges obtained from those suggested by the Health Care Finance Administration as acceptable performance criteria for deviation from a target value
  • Due to method differences and matrix interactions, the values listed may not always be reproduced by other analytical systems
  • Each laboratory should establish its own precision parameters
Specifications
Specifications
2 x 1 mL
CE
Glycohemoglobin Control Set
Lyophilized
In-vitro diagnostic use, quantitative determination of Human glycohemoglobin (HbA1) in Blood by cation exchange resin