Pointe Scientific General Chemistry Standards

Manufacturer: Pointe Scientific G7540STD

 View more versions of this product

Catalog No. 23-666-298

  Check Availability
Add to Cart

    Specifications

    Product Type Glycohemoglobin Standard
    Format Lyophilized
    Includes Approximately 10%
    For Use With (Application) For the purpose of standardizing results obtained from run to run in the quantitative determination of Human Glycohemoglobin (HbA1) in Blood by cation exchange Resin
    Quantity 1 x 1mL
    Certifications/Compliance CE
    Color Dark Red
    Stability 1 Month
    Concentration 10%
    Storage Requirements 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit