Pointe Scientific™ Glucose Dry Powder Reagents

Quantify glucose levels in serum samples

Manufacturer: Pointe Scientific G75191L

 View more versions of this product

Catalog No. 23-666-289

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Glucose (Oxidase) reagent set
    For Use With (Application) Quantitative determination of Glucose in Serum, in-vitro diagnostic use only
    Format Dry powder
    pH 7.4
    Quantity 200mL
    Certifications/Compliance CE Marked
    Formulation Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit