Pointe Scientific Glucose (Hexokinase) Reagents

For the quantitative determination of Glucose in serum

Manufacturer: Pointe Scientific G7518400

 View more versions of this product

Catalog No. 23-666-285

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Glucose in Serum
    Format Dry powder
    Formulation Hexokinase 1000U/L, G6PDH 1000U/L, ATP 1mM, NAD 1mM, Buffer pH 7.5 ± 0.1, Sodium Azide 0.01% w/v, non-reactive stablilizers and fillers
    pH 7.5
    Quantity 8 × 50mL
    Product Type Glucose (Hexokinase) reagent

    • Concentration: 65-110mg/dL
    • Reconstituted Stability: 5 days
    • Linearity: 600mg/dL
    • Storage temperature: 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit