Pointe Scientific™ γ-Glutamyl Transferase Reagents for Hitachi™ Analyzers

Quantify γ-GT easily with system specific reagents

Manufacturer: Pointe ScientificSupplier Diversity Partner HG759140

 View all options for this product

Catalog No. 23-666-259

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum
    For Use With (Equipment) Hitachi™ 704, 902 Analyzer
    Format Liquid
    Formulation R1 and R2 contains Tris Buffer 89mM/L pH 8.1 ±0.1, Glycylglycine 126mM/L, GLUPA-C 3.2mM/L, Sodium Azide 0.095%
    Quantity 5 x 20mL R1, 2 x 20mL R2
    Size 5 × 20mL
    Product Type gamma-Glutamyl Transferase reagent kit

    • Method: Modified Szasz, Kinetic
    • Format: Liquid (ready to use)
    • Linearity: 800 U/L
    • Expected Values: 6 to 54 U/L

    Applications:

    For the quantitative kinetic determination of γ-glutamyl transferase (γ-GT) activity in serum on Hitachi analyzers

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit