Learn More
Pointe Scientific Glycohemoglobin Reagents
Manufacturer:  Pointe Scientific G754080
 View more versions of this product
Catalog No. 23-666-294
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CE Marked
Liquid
1 x 120mL Resin, 1 x 30mL Lysing, 80 Serum separators
Quantitative determination of Glycohemoglobin (HbA1) in Blood by cation exchange resin, test is to be used to monitor long-term Glucose control in diabetes mellitus
1 x 120mL Bottle 8mg/mL cation-exchange resin in a Borate Buffer, pH 6.9, 1 x 30mL bottle Glycohemoglobin Lysing resin, 10mM Potassium Cyanide, Surfactant added, 40 Serum separators
Glycohemoglobin reagent