Pointe Scientific HDL Cholesterol Reagents

Manufacturer: Pointe Scientific H750780

 View more versions of this product

Catalog No. 23-666-299

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type HDL Cholesterol precipitating reagent set- Mg/Dex
    Format Dry powder
    For Use With (Application) Quantitative determination of High Density Lipoprotein in Serum
    Formulation HDL Cholesterol precipitating reagent: Dextran Sulfate (50000 MW) 10g/L, Magnesium ions 500mM, Buffer pH 7 ±0.1, non-reactive ingredients with 0.1% Sodium Azide as preservative
    Quantity 8 x 10mL
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit