Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Iron-Total Reagents for Hitachi Analyzers
Quantify iron levels in serum
Supplier: Pointe Scientific HI904600
Includes: Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5) with surfactant
Color Reagent: Ferrozine 3.6mM in hydroxylamine hydrochloride
Requires : Requires calibrator/standard
This item is not returnable. View return policy
Description
- Method: Ferrozine
- Format: Liquid (ready to use)
- Linearity: 500ug/dL
- Expected Values: 60 to 150ug/dL
Applications:
For the quantitative determination of iron in serum on Hitachi analyzers
Specifications
CE Marked | |
Total Iron Reagent Kit | |
Quantitative determination of Iron in Serum |
Liquid | |
Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, Iron color R2: 3.6mM Ferrozine in Hydroxylamine Hydrochloride | |
5 x 100mL R1, 1 x 100mL R2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostics use only