Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE 1N KCL Solution Reagents

Specifications

Certifications/Compliance CE Marked
Format Liquid
Type 1N KCL Solution Reagent Kit
Formulation Potassium Chloride 1mol/L, Anti-microbial 0.01%
For Use With (Application) Quantitative determination of Sodium, Potassium and Chloride in Serum
View More Specs

Products 2
Catalog Number Mfr. No. Quantity ForUseWithEquipment Price Quantity & Availability  
Catalog Number Mfr. No. Quantity ForUseWithEquipment Price Quantity & Availability  
23-666-325
View Documents
Pointe Scientific
HI790KCL
1 x 1 L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
23-666-322
View Documents
Pointe Scientific
HI990KCL
5 x 300 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

CE Marked
1N KCL Solution Reagent Kit
Quantitative determination of Sodium, Potassium and Chloride in Serum
Liquid
Potassium Chloride 1mol/L, Anti-microbial 0.01%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit