Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific ISE Solution Reagents for Roche Cobas Analyzers
Needed for quantification of sodium, potassium and chloride on Roche Cobas systems
Catalog Number | Mfr. No. | Quantity | Formulation | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Formulation | Product Type | Price | Quantity & Availability | |||
23-666-448
|
Pointe Scientific
MIR160 |
60mL | KCl aqueous solution 3.5mM/L | ISE solution reagent |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-447
|
Pointe Scientific
MES525 |
125mL | Ammonium Hydrogen Difluoride 0.6% | ISE solution reagent |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-449
|
Pointe Scientific
MIS601 |
250mL | Aqueous solution containing: Na+ 150mM/L, K+ 3mM/L, Cl 138mM/L and a preservative | ISE standard 1 solution reagent |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-450
|
Pointe Scientific
MIS602 |
60mL | Aqueous solution containing: Na+ 100mM/L, K+ 7mM/L, Cl- 92mM/L and a preservative | ISE standard 2 solution reagent |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Method: Ion-selective Electrode (ISE)
- Format: Liquid
- Reference Solution contains: KCl 3.5mM
- Standard Solution 1 contains: Na+ 150mM; K+ 3mM; Cl- 138mM; Preservative
- Standard Solution 2 contains: Na+ 100mM; K+ 7mM; Cl- 92mM; Preservative
- Etcher Solution contains: Ammonium hydrogen difluoride 0.6%
For ISE applications on Roche Cobas Chemistry Systems
Specifications
Cobas™ Mira™ analyzer |
For in vitro diagnostic use only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title