Pointe Scientific™ Lactate Dehydrogenase Dry Powder Reagents

Quantify lactate dehydrogenase activity in serum

Manufacturer: Pointe Scientific L7535300

 View more versions of this product

Catalog No. 23-666-352

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Disclaimers:

For in vitro diagnostic use only


    Specifications

    Product Type Lactate Dehydrogenase reagent set
    For Use With (Application) Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum, in-vitro diagnostic use only
    Format Dry powder
    Quantity 15mL
    Size 20 × 15mL
    Wavelength 340nm
    Certifications/Compliance CE Marked
    Formulation NAD 7mM, L-Lactate 50mM, Tris Buffer pH 9 ± 0.1, non-reactive Stabilizers and fillers
    pH 9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit