Learn More
Pointe Scientific™ Lactate Dehydrogenase Dry Powder Reagents
Quantify lactate dehydrogenase activity in serum
Manufacturer:  Pointe Scientific L7535-400
 View more versions of this product
Catalog No. 23-666-355
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Disclaimers:
For in vitro diagnostic use only
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
9
8 × 50mL
Lactate Dehydrogenase reagent set
Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum, in-vitro diagnostic use only
NAD 7mM, L-Lactate 50mM, Tris Buffer pH 9 ± 0.1, non-reactive Stabilizers and fillers
50mL
340nm