Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Lactate Dehydrogenase Reagents for Hitachi Analyzers
Quantify lactate dehydrogenase activity easily with system specific reagents
Specifications
Certifications/Compliance | CE Marked |
---|---|
Format | Liquid |
Includes: Reagent 1 and Reagent 2 when combined: NAD 7.5mM; L-Lactate 55mM; Buffer (pH 8.95); Non-reactive stabilizers; Sodium azide 0.1%
Catalog Number | Mfr. No. | Quantity | Size | Formulation | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Size | Formulation | Price | Quantity & Availability | |||
23-666-344
|
Pointe Scientific
HL456R2 |
5 x 100 R2mL | Sodium Azide 0.095% |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-343
|
Pointe Scientific
HL456R1 |
5 x 600 R1mL | 5 × 600mL | Sodium Azide 0.095% |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-346
|
Pointe Scientific
HL756284 |
4 x 50mL R1, 4 x 21mL R2 | 4 × 50mL | NAD 7.5mM, L-Lactate 55mM, Buffer pH 8.95, non-reactive Stabilizers and Sodium Azide 0.1% as preservative |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-666-347
|
Pointe Scientific
HL956568 |
4 x 100, 8 x 21mL | 4 × 100mL | Sodium Azide 0.095% |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Method: Modified Wacker/Tris, Kinetic
- Format: Liquid (ready to use)
- Linearity: 1000 U/L
- Expected Values: 50 to 166 U/L (Male, 30°C); 60 to 132 U/L (Female, 30°C)
Applications:
For the quantitative kinetic determination of lactate dehydrogenase activity in serum on Hitachi analyzers
Specifications
CE Marked | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title