Learn More
Pointe Scientific™ Lipase Dry Powder Reagents
Quantify pancreatic lipase levels in serum
Manufacturer:  Pointe Scientific L7503100
 View more versions of this product
Catalog No. 23-666-357
  Check Availability
Add to cart
Disclaimers:
For in vitro diagnostic use only
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
Lipase Substrate and Buffer reagents, Lipase activator solution, Lipase standard
10 × 10mL
Lipase reagent set
Quantitative kinetic determination of Pancreatic Lipase in Serum
Lipase Substrate contain: 1,2-Diglyceride 63%, MGLP 87U/100mL, GK 133U/100mL, GPO 4000U/100mL, Co-Lipase 4000U/100mL, Buffer, Substrate Buffer and Activator contains Sodium Azide 0.05%, Standard contains Caution contains Human source materials
10 x 10mL Substrate, 1 x 105mL Substrate Buffer, 1 x 30mL Activator, 1 x 3mL Standard
550nm