Pointe Scientific™ Lipase Dry Powder Reagents

Quantify pancreatic lipase levels in serum

Manufacturer: Pointe Scientific L750340

 View more versions of this product

Catalog No. 23-666-358

  Check Availability
Add to Cart

Disclaimers:

For in vitro diagnostic use only


    Specifications

    Product Type Lipase reagent set
    Formulation Lipase Substrate contain: 1,2-Diglyceride 63%, MGLP 87U/100mL, GK 133U/100mL, GPO 4000U/100mL, Co-Lipase 4000U/100mL, Buffer, Substrate Buffer and Activator contains Sodium Azide 0.05%, Standard contains Caution contains Human source materials
    Quantity 4 x 10mL Substrate, 2 x 21mL Substrate Buffer, 1 x 14mL Activator, 1 x 3mL Standard
    Size 4 × 10mL
    For Use With (Application) Quantitative kinetic determination of Pancreatic Lipase in Serum
    Format Dry powder
    Includes Lipase Substrate and Buffer reagents, Lipase activator solution, Lipase standard
    Wavelength 550nm
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit