Pointe Scientific™ Microprotein Reagents

Quantify total protein levels in urine

Manufacturer: Pointe ScientificSupplier Diversity Partner HP782-200

 View all options for this product

Catalog No. 23-666-374


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of total protein in urine
    For Use With (Equipment) Hitachi™ 704, 902 analyzer
    Format Liquid
    Formulation Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants, preservative
    Model 704, 902
    Quantity 4 x 50mL Reagent, 1 x 15mL Standard
    Size 4 × 50mL (1.7 oz.)
    Product Type Microprotein reagent kit

    • Method: Pyragallol Red, Endpoint
    • Format: Liquid (ready to use)
    • Wavelength: 600nm
    • Linearity: 250mg/dL
    • Expected Values: 28 to 141mg (24 hr.)

    For the quantitative determination of total protein in urine

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit