Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Phosphorus Reagents for Hitachi™ Analyzers
Quantify inorganic phosphorus easily with system specific reagents
Supplier: Pointe Scientific HP9161200
Includes: Reagent 1 and Reagent 2 when combined: Ammonium molybdate >0.48mM; Sulfuric acid <220mM; surfactant
Requires : Calibrator/standard
This item is not returnable. View return policy
Description
- Method: Phosphomolybdate, Endpoint
- Format: Liquid (ready to use)
- Linearity: 12mg/dL
- Expected Values: 2.5 to 4.8mg/dL
Applications:
For the quantitative determination of inorganic phosphorus in serum on Hitachi analyzers
Specifications
CE Marked | |
Ammonium Molybdate >0.48mM, Sulfuric Acid <220mM with Surfactant | |
8 x 100mL R1, 4 x 100mL R2 |
Phosphorus Reagent Kit | |
Quantitative determination of inorganic Phosphorus in Serum | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction