Pointe Scientific™ Protein-Total Reagents

Quantify total protein in serum

Manufacturer: Pointe Scientific HT9281200

 View more versions of this product

Catalog No. 23-666-408

  Check Availability
Add to Cart

Disclaimers:

For in vitro diagnostic use only


    Specifications

    Product Type Total Protein reagent kit
    For Use With (Application) Quantitative determination of total Protein concentration in Serum, in-vitro diagnostic use only
    For Use With (Equipment) Hitachi™ 717, 911, 912, 914 analyzer
    Format Liquid
    Formulation R1 and R2 contains Sodium Hydroxide 750mM, Copper Sulfate 6mM, Sodium Potassium Tartrate 35mM, Potassium Iodide 6mM, non-reactive ingredients
    Model 717, 911, 912, 914
    pH 13.2
    Quantity 100mL
    Size 8 × 100mL (3.4 oz.)
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit