Pointe Scientific™ Total Bilirubin Reagents

For quantifying total bilirubin

Manufacturer: Pointe Scientific B7576-500

 View more versions of this product

Catalog No. 23-666-152

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Total Bilirubin reagent kit
    For Use With (Application) Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only
    Formulation Total Bilirubin reagent: Sulfanilic Acid 16mM, Hydrochloric Acid 164mM, Dimethyl Sulfoxide 4.4M, Surfactants, Sodium Nitrite 0.6M
    Quantity 500mL
    Size 1 × 500mL
    Format Liquid
    Wavelength 550nm
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit