Pointe Scientific™ Triglycerides Dry Powder Reagents

Quantify triglyceride levels in serum or plasma

Manufacturer: Pointe Scientific T75311L

 View more versions of this product

Catalog No. 23-666-416

  Check Availability
Add to Cart

    Specifications

    Product Type Triglyceride reagent set
    For Use With (Application) Quantitative determination of Triglyceride in Serum or Plasma, in-vitro diagnostic use only
    Format Dry powder
    Formulation Triglyceride reagent (Concentrations refer to reconstituted reagent) ATP 1mM, Magnesium Salt >5mM, TBHB 2mM, GPO >2000U/L, Lipase >200000U/L, GK 1,000U/L, Peroxidase >500U/L, Buffer, Surfactants, Stabilizers, Fillers, Sodium Azide 0.1%
    pH 6.5
    Quantity 250mL
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit