Learn More
Pointe Scientific™ Triglycerides Dry Powder Reagents
Quantify triglyceride levels in serum or plasma
Manufacturer:  Pointe Scientific T7531400
 View more versions of this product
Catalog No. 23-666-418
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
6.5
Triglyceride reagent set
Quantitative determination of Triglyceride in Serum or Plasma, in-vitro diagnostic use only
Triglyceride reagent (Concentrations refer to reconstituted reagent) ATP 1mM, Magnesium Salt >5mM, TBHB 2mM, GPO >2000U/L, Lipase >200000U/L, GK 1,000U/L, Peroxidase >500U/L, Buffer, Surfactants, Stabilizers, Fillers, Sodium Azide 0.1%
50mL