Pointe Scientific™ Uric Acid Dry Powder Reagents

Quantify uric acid levels in serum

Manufacturer: Pointe ScientificSupplier Diversity Partner U7580-400

 View all options for this product

Catalog No. 23-666-437


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Uric Acid in Serum
    Format Powder
    Formulation 4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
    Quantity 8 x 50mL
    Product Type Uric Acid standard reagent

    • Method: Uricase/Trinder, Endpoint
    • Format: Dry powder
    • Wavelength: 520nm
    • Linearity: 25mg/dL
    • Expected Values: 2.5 to 7.7mg/dL
    • Reconstituted stability: 48 hours RT, 31 days 2 to 8°C

    For the quantitative determination of Uric Acid in serum

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit