Pointe Scientific Uric Acid Liquid Reagents

Reagent requires Uric Acid Standard (23-666-424)/Chemistry Calibrator (23-666-189)

Manufacturer: Pointe ScientificSupplier Diversity Partner U7580-150

 View all options for this product

Catalog No. 23-666-433


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance CE Marked
    For Use With (Application) Quantitative determination of Uric Acid in Serum, in-vitro diagnostic use only
    Format Dry powder
    Formulation 4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
    pH 8.1
    Quantity 10 x 15mL
    Product Type Uric Acid reagent set

    Method: Uricase/trinder, endpoint
    Storage Temperature: 2–8°C
    Reconstituted Stability: Ready-to-use liquid
    Linearity: 20mg/dL
    Measurement Wavelength: 520nm
    Reaction Temperature: 37°C
    Expected Values: 2.5–7.7mg/dL

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit