Learn More
Pointe Scientific Uric Acid Liquid Reagents
Reagent requires Uric Acid Standard (23-666-424)/Chemistry Calibrator (23-666-189)
Manufacturer:  Pointe Scientific U7580300
 View more versions of this product
Catalog No. 23-666-434
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
CE Marked
Dry powder
8.1
Uric Acid reagent set
Quantitative determination of Uric Acid in Serum, in-vitro diagnostic use only
4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
20 x 15mL